Human recombinant siglec-3 (from HEK293 cells), FITC (fluorescein isothiocyanate)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
FITC-Labeled Human Siglec-3 / CD33 Protein, Fc Tag.
Specifications
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag
- Storage conditions:–20 ℃, aviod light
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Gene ID:P20138-1 (R69G)
- Protein synonyms:gp67|SIGLEC3|CD33
- Protein/peptide name:Siglec-3
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:52,9 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT