Human recombinant nectin-4 (from HEK293 cells), Biotin
Biotinylated Human Nectin-4 Protein, His,Avitag™.
- Conjugation:Biotin
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His, Avitag™
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilised Biotinylated Human Nectin-4, His,Avitag (Cat. No. NE4-H82E7) at 1 μg/ml (100 μl/well) on streptavidin precoated (0,5 μg/well) plate, can bind Human Nectin-1, Fc Tag (Cat. No. PV1-H5253) with a linear range of 2 - 40 ng/ml (QC tested).
- Gene ID:Q96NY8-1
- Protein synonyms:PVRL4|NECTIN4| PRR4|LNIR
- Protein/peptide name:Nectin-4
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:37,8 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:RT