Mouse recombinant VEGF R1 (from HEK293)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:−20 °C
- Endotoxin content:Less than 0,1 EU per μg by the LAL method
- Biological activity:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM glycine, pH 7,5. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised human VEGF165, Tag Free (Cat. No. ACRBVE5-H4210) at 2 μg/ml (100 μl/well) can bind mouse VEGF R1, mouse IgG2a Fc Tag, low endotoxin (Cat. No. ACRBVE1-M5256) with a linear range of 2 - 31 ng/ml (QC tested).; Measured by its binding ability in a functional ELISA. Immobilised human VEGF-B, His Tag (Cat. No. ACRBVE6-H5225) at 2 μg/ml (100 μl/well) can bind mouse VEGF R1, mouse IgG2a Fc Tag, low endotoxin (Cat. No. ACRBVE1-M5256) with a linear range of 8 - 63 ng/ml (Routinely tested).; Measured by its binding ability in a functional ELISA. Immobilised mouse VEGF120, Tag Free (Cat. No. ACRBVE0-M4211) at 2 μg/ml (100 μl/well) can bind mouse VEGF R1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. ACRBVE1-M5256) with a linear range of 4 - 63 ng/ml (Routinely tested).; Measured by its binding ability in a functional ELISA. Immobilised mouse VEGF164, Tag Free (Cat. No. ACRBVE4-M4216) at 2 μg/ml (100 μl/well) can bind mouse VEGF R1, mouse IgG2a Fc Tag, low endotoxin (Cat. No. ACRBVE1-M5256) with a linear range of 2 - 31 ng/ml (Routinely tested).
- Gene ID:P35969-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:FLT|FLT1|VEGFR1
- Protein/peptide name:VEGF R1
- Purity:>85%
- Molecular weight:109,8 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM glycine, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT