Order Entry
Germany
ContactUsLinkComponent
Human recombinant GUCY2C (from HEK293 cells)
Human recombinant GUCY2C (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant GUCY2C (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    2e5 of anti-GUCY2C CAR-293 cells were stained with 100 µl of 1 µg/ml Human GUCY2C, Fc Tag (Cat. No. GUC-H5257) and negative control protein respectively, washed and then followed by PE anti-human IgG Fc and analysed with FACS (Routinely tested).
  • Gene ID:
    P25092-1
  • Protein synonyms:
    GUCY2C|hSTAR|STA receptor|GC-C|STAR|GUC2C
  • Protein/peptide name:
    GUCY2C
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    72,5 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications