Human recombinant IL-6 R alpha (from HEK293)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Human IL-6, Tag Free (Cat. No. ACRBIL6-H4218) at 5 μg/ml (100 μl/well) can bind Human IL-6 R alpha, Fc Tag (Cat. No. ACRBILR-H5259) with a linear range of 1 - 20 ng/ml (QC tested).
- Gene ID:P08887-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:IL-6RA|IL6RA|IL-6R-alpha|IL6RQ|IL-6R-1|IL6R|CD126|gp80
- Protein/peptide name:IL-6 R alpha
- Purity:>95%
- Molecular weight:65,0 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT