Order Entry
Germany
ContactUsLinkComponent
 
Human recombinant Gremlin (from HEK293), Acro Biosystems
 
Human recombinant Gremlin (from HEK293), Acro Biosystems

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4 with 1 mM EDTA. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Human BMP-2, Tag Free (Cat. No. ACRBBM2-H4117) at 2 μg/ml (100 μl/well) can bind Human Gremlin, His Tag (Cat. No. ACRBGR1-H52H3) with a linear range of 0,02 - 0,313 μg/ml (QC tested).
  • Gene ID:
    O60565
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    CKTSF1B1|DRM|DAND2|PIG2|GREM1
  • Protein/peptide name:
    Gremlin
  • Purity:
    >95%
  • Molecular weight:
    20,2 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4 with 1 mM EDTA. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT