Order Entry
Germany
ContactUsLinkComponent
 
Rat recombinant PD-1 (from HEK293)
Catalog # ACRBPD1-R52H2-1MG
Supplier:  ACROBIOSYSTEMS
CAS Number:  
Rat recombinant PD-1 (from HEK293)
Catalog # ACRBPD1-R52H2-1MG
Supplier:  ACROBIOSYSTEMS
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Rat
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised Rat PD-1, His Tag (Cat. No. PD1-R52H2) at 2 μg/ml (100 μl/well) can bind Mouse PD-L1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. PD1-M52A2) with a linear range of 0,02-0,313 μg/ml (QC tested).
  • Gene ID:
    D3ZIN8
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    CD279|PDCD1|PD1|SLEB2
  • Protein/peptide name:
    PD-1
  • Purity:
    >95%
  • Molecular weight:
    18,1 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

About this item