Order Entry
Germany
ContactUsLinkComponent
Racks for disposal bags
Racks for disposal bags
Catalog # 129-0972
Supplier:  Brand
undefined
Racks for disposal bags
Catalog # 129-0972
Supplier:  Brand

Specifications

  • Description:
    Stand for disposal bags, white
  • Seq:
    05
  • Pk:
    1

Specifications

About this item

These bags are made of epoxy-coated steel wire, with rubber feet.

  • Not autoclavable
  • Inner diameter: 120 mm
  • Height: 250 mm