Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,1-Difluoro-3-(iodomethyl)cyclobutane
1,1-Difluoro-3-(iodomethyl)cyclobutane
1,1-Difluoro-3-(iodomethyl)cyclobutane
1,1-Difluoro-3-(iodomethyl)cyclobutane
Supplier:
Apollo Scientific
Certificates
Expand image
1,1-Difluoro-3-(iodomethyl)cyclobutane
Supplier:
Apollo Scientific
About this item
CAS Number:
1434142-22-9
:
Formula:
C₅H₇F₂I
MDL Number:
MFCD26521335
:
232.01 g/mol
Storage Temperature:
Ambient