2-(2-Fluorophenyl)-2-methylpropan-1-ol 95%
Catalog # APOSPC907490-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:168.208 g/mol
Storage Temperature:Ambient
CAS Number:1177192-02-7
Formula:C₁₀H₁₃FO
MDL Number:MFCD11934574