Order Entry
Germany
ContactUsLinkComponent
2,4-Dichloro-6-fluorobenzonitrile 95%
2,4-Dichloro-6-fluorobenzonitrile 95%
Catalog # APOSPC906758-1G
Supplier:  Apollo Scientific
CAS Number:  
2,4-Dichloro-6-fluorobenzonitrile 95%
Catalog # APOSPC906758-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :190.002 g/mol
Storage Temperature:Ambient
CAS Number:1349718-98-4
Formula:C₇H₂Cl₂FN
MDL Number:MFCD19686188