Order Entry
Germany
ContactUsLinkComponent
3-Hydroxy-4-(trifluoromethyl)benzonitrile 95%
3-Hydroxy-4-(trifluoromethyl)benzonitrile 95%
Catalog # APOSPC906689-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Hydroxy-4-(trifluoromethyl)benzonitrile 95%
Catalog # APOSPC906689-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :187.119 g/mol
Storage Temperature:Ambient
CAS Number:731002-50-9
Formula:C₈H₄F₃NO
MDL Number:MFCD16999170