Order Entry
Germany
ContactUsLinkComponent
2-Fluoro-5-hydroxy-3-methoxypyridine 95%
2-Fluoro-5-hydroxy-3-methoxypyridine 95%
Catalog # APOSPC905174-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Fluoro-5-hydroxy-3-methoxypyridine 95%
Catalog # APOSPC905174-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :143.116 g/mol
Storage Temperature:Ambient
CAS Number:1227511-69-4
Formula:C₆H₆FNO₂
MDL Number:MFCD16611061