Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(1Z)-1,3,3,3-Tetrafluoroprop-1-ene
(1Z)-1,3,3,3-Tetrafluoroprop-1-ene
(1Z)-1,3,3,3-Tetrafluoroprop-1-ene
:
Apollo Scientific
(1Z)-1,3,3,3-Tetrafluoroprop-1-ene
:
Apollo Scientific
:
29118-25-0
:
:
C₃H₂F₄
:
MFCD18071081
:
114.042 g/mol
:
21 °C
:
3161
:
2.1,
:
GHS02
:
GHS04
:
Ambient