Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Acetoxy-2,3,5,6-tetrafluorobenzoic acid
4-Acetoxy-2,3,5,6-tetrafluorobenzoic acid
4-Acetoxy-2,3,5,6-tetrafluorobenzoic acid
APOSPC5714-250MG
:
Apollo Scientific
:
4-Acetoxy-2,3,5,6-tetrafluorobenzoic acid
APOSPC5714-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
252.119 g/mol
:
GHS07
:
Ambient
:
83789-90-6
:
C₉H₄F₄O₄
:
MFCD03094301