Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-2,4,5-trifluoroacetopheone
3-Bromo-2,4,5-trifluoroacetopheone 95%
3-Bromo-2,4,5-trifluoroacetopheone 95%
:
Apollo Scientific
3-Bromo-2,4,5-trifluoroacetopheone 95%
:
Apollo Scientific
:
1896891-81-8
:
:
C₈H₄BrF₃O
:
MFCD31568658
:
253.016 g/mol
:
GHS07
:
Ambient