Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Fluorobenzo[b]furan
7-Fluorobenzo[b]furan
7-Fluorobenzo[b]furan
Irritant
7-Fluorobenzo[b]furan
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
7-Fluorobenzo[b]furan
Supplier:
Apollo Scientific
About this item
CAS Number:
24410-61-5
:
Formula:
C₈H₅FO
MDL Number:
MFCD15143817
:
136.123 g/mol
:
GHS07
Storage Temperature:
Ambient