Order Entry
Germany
ContactUsLinkComponent
 
N-Boc-(S)-2,4,5-trifluoro-b-homophenylalanine 97%
Catalog # APOSPC530015-1G
Supplier:  Apollo Scientific
CAS Number:  
N-Boc-(S)-2,4,5-trifluoro-b-homophenylalanine 97%
Catalog # APOSPC530015-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :333.303 g/mol
Storage Temperature:Ambient
CAS Number:922178-94-7
Formula:C₁₅H₁₈F₃NO₄
MDL Number:MFCD06659150