Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Fluoro-6-methoxybenzenesulphonyl chloride
2-Fluoro-6-methoxybenzenesulphonyl chloride, tech
2-Fluoro-6-methoxybenzenesulphonyl chloride, tech
2-Fluoro-6-methoxybenzenesulphonyl chloride, tech
Supplier:
Apollo Scientific
Certificates
Expand image
2-Fluoro-6-methoxybenzenesulphonyl chloride, tech
Supplier:
Apollo Scientific
About this item
CAS Number:
1176126-31-0
:
Formula:
C₇H₆ClFO₃S
MDL Number:
MFCD12922637
:
224.637 g/mol
Storage Temperature:
Ambient
UN:
3261
ADR:
8,II
:
GHS05