Order Entry
Germany
ContactUsLinkComponent
4-(Difluoromethyl)-1,5-dimethoxy-naphthalene
4-(Difluoromethyl)-1,5-dimethoxy-naphthalene
Catalog # APOSPC520350-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(Difluoromethyl)-1,5-dimethoxy-naphthalene
Catalog # APOSPC520350-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :238.23 g/mol
Storage Temperature:Ambient
CAS Number:2149597-07-7
Formula:C₁₃H₁₂F₂O₂
MDL Number:MFCD29767792