Order Entry
Germany
ContactUsLinkComponent
6-(Difluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carbonitrile
6-(Difluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carbonitrile
Catalog # APOSPC520057-1G
Supplier:  Apollo Scientific
CAS Number:  
6-(Difluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carbonitrile
Catalog # APOSPC520057-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :194.141 g/mol
Storage Temperature:Ambient
Formula:C₈H₄F₂N₄
MDL Number:MFCD29767540