Ethyl 5-amino-4-bromo-1-[4-(trifluoromethoxy)phenyl]pyrazole-3-carboxylate
Catalog # APOSPC520016-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:394.144 g/mol
Storage Temperature:Ambient
CAS Number:1427021-51-9
Formula:C₁₃H₁₁BrF₃N₃O₃
MDL Number:MFCD23135145