Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2-(difluoromethyl)isonicotinate
Methyl 2-(difluoromethyl)isonicotinate
Methyl 2-(difluoromethyl)isonicotinate
Irritant
Methyl 2-(difluoromethyl)isonicotinate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl 2-(difluoromethyl)isonicotinate
Supplier:
Apollo Scientific
About this item
CAS Number:
1251844-44-6
:
Formula:
C₈H₇F₂NO₂
MDL Number:
MFCD22690248
:
187.143 g/mol
:
GHS07
Storage Temperature:
Ambient