Order Entry
Germany
ContactUsLinkComponent
 
Irritant
2-Fluoro-3-hydroxy-5-methylpyridine
Catalog # APOSPC51125-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Fluoro-3-hydroxy-5-methylpyridine
Catalog # APOSPC51125-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :127.116 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1184172-53-9
Formula:C₆H₆FNO
MDL Number:MFCD16611070