Ethyl 6-Fluoro-1,2,3,4-tetrahydro-isoquinoline-1-carboxylate hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1822454-85-2
:
Formula:C₁₂H₁₄FNO₂·ClH
MDL Number:MFCD28155351
:259.704 g/mol
Storage Temperature:Ambient