0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
2-Hydroxy-4-(trifluoromethyl)benzaldehyde
2-Hydroxy-4-(trifluoromethyl)benzaldehyde 97%
2-Hydroxy-4-(trifluoromethyl)benzaldehyde 97%
Irritant
2-Hydroxy-4-(trifluoromethyl)benzaldehyde 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Hydroxy-4-(trifluoromethyl)benzaldehyde 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
58914-34-4
:
Formula:
C₈H₅F₃O₂
MDL Number:
MFCD04039240
:
190.119 g/mol
:
GHS07
Storage Temperature:
Ambient