Order Entry
Germany
ContactUsLinkComponent
Methyl 1-Methyl-5-[4-(trifluoromethyl)phenyl]pyrrole-2-carboxylate
Methyl 1-Methyl-5-[4-(trifluoromethyl)phenyl]pyrrole-2-carboxylate
Catalog # APOSPC510247-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 1-Methyl-5-[4-(trifluoromethyl)phenyl]pyrrole-2-carboxylate
Catalog # APOSPC510247-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :283.246 g/mol
Storage Temperature:Ambient
CAS Number:1895780-61-6
Formula:C₁₄H₁₂F₃NO₂
MDL Number:MFCD28954611