Methyl 4-Oxo-1-(2,4,5-trifluorophenyl)cyclohexanecarboxylate
Catalog # APOSPC510129-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:286.246 g/mol
Storage Temperature:Ambient
CAS Number:1385694-46-1
Formula:C₁₄H₁₃F₃O₃
MDL Number:MFCD22423134