Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-2,4,5-trifluorobenzenboronic acid
3-Bromo-2,4,5-trifluorobenzenboronic acid 95%
3-Bromo-2,4,5-trifluorobenzenboronic acid 95%
3-Bromo-2,4,5-trifluorobenzenboronic acid 95%
Supplier:
Apollo Scientific
Certificates
Expand image
3-Bromo-2,4,5-trifluorobenzenboronic acid 95%
Supplier:
Apollo Scientific
About this item
:
Formula:
C₆H₃BBrF₃O₂
MDL Number:
MFCD32662413
:
254.797 g/mol
Storage Temperature:
Refrigerator