Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-2,4,5-trifluorobenzaldehyde
3-Bromo-2,4,5-trifluorobenzaldehyde 95%
3-Bromo-2,4,5-trifluorobenzaldehyde 95%
Irritant
3-Bromo-2,4,5-trifluorobenzaldehyde 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Bromo-2,4,5-trifluorobenzaldehyde 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1260903-08-9
:
Formula:
C₇H₂BrF₃O
MDL Number:
MFCD12756731
:
238.989 g/mol
:
GHS07
Storage Temperature:
Ambient