Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3-bromo-2,4,5-trifluorobenzoate
Methyl 3-bromo-2,4,5-trifluorobenzoate 95+%
Methyl 3-bromo-2,4,5-trifluorobenzoate 95+%
Irritant
Methyl 3-bromo-2,4,5-trifluorobenzoate 95+%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl 3-bromo-2,4,5-trifluorobenzoate 95+%
Supplier:
Apollo Scientific
About this item
CAS Number:
195532-60-6
:
Formula:
C₈H₄BrF₃O₂
MDL Number:
MFCD11500619
:
269.015 g/mol
:
GHS07
Storage Temperature:
Ambient