Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-(Trifluoromethyl)pyridine-2-carboxaldehyde
6-(Trifluoromethyl)pyridine-2-carboxaldehyde 99%
6-(Trifluoromethyl)pyridine-2-carboxaldehyde 99%
Irritant
6-(Trifluoromethyl)pyridine-2-carboxaldehyde 99%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
6-(Trifluoromethyl)pyridine-2-carboxaldehyde 99%
Supplier:
Apollo Scientific
About this item
CAS Number:
131747-65-4
:
Formula:
C₇H₄F₃NO
MDL Number:
MFCD10696233
:
175.108 g/mol
:
GHS07
Storage Temperature:
Refrigerator