Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-3-(trifluoromethyl)phenol
2-Bromo-3-(trifluoromethyl)phenol
2-Bromo-3-(trifluoromethyl)phenol
2-Bromo-3-(trifluoromethyl)phenol
Supplier:
Apollo Scientific
Certificates
Expand image
2-Bromo-3-(trifluoromethyl)phenol
Supplier:
Apollo Scientific
About this item
CAS Number:
1214323-39-3
:
Formula:
C₇H₄BrF₃O
MDL Number:
MFCD13194487
:
241.005 g/mol
Storage Temperature:
Ambient