
Methyl 4-fluoro-2-(methoxymethyl)-5-nitrobenzoate
Catalog # APOSPC49238-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:243.189 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1243164-13-7
Formula:C₁₀H₁₀FNO₅
MDL Number:MFCD22378816