Order Entry
Germany
ContactUsLinkComponent
(1-Ethoxy-2,2,2-trifluoroethoxy)trimethylsilane
(1-Ethoxy-2,2,2-trifluoroethoxy)trimethylsilane
Catalog # APOSPC450525-1G
Supplier:  Apollo Scientific
CAS Number:  
(1-Ethoxy-2,2,2-trifluoroethoxy)trimethylsilane
Catalog # APOSPC450525-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :216.274 g/mol
Boiling Pt:55…56 °C (60 mm Hg)
Storage Temperature:Ambient
CAS Number:141023-08-7
Formula:C₇H₁₅F₃O₂Si
MDL Number:MFCD30730308