(1-Ethoxy-2,2-difluoroethoxy) triethylsilane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1922958-24-4
:
Formula:C₁₀H₂₂F₂O₂Si
MDL Number:MFCD30730298
:240.363 g/mol
Boiling Pt:69…70 °C (6 mm Hg)
Storage Temperature:Ambient