sec-Butyl hexafluoroisopropyl carbonate
Catalog # APOSPC450483-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:268.154 g/mol
Storage Temperature:Ambient
CAS Number:1923065-41-1
Formula:C₈H₁₀F₆O₃
MDL Number:MFCD28334454