Order Entry
Germany
ContactUsLinkComponent
4-Bromo-3,3,4,4-tetrafluorobutyl sec-butyl carbonate
4-Bromo-3,3,4,4-tetrafluorobutyl sec-butyl carbonate
Catalog # APOSPC450482-25G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-3,3,4,4-tetrafluorobutyl sec-butyl carbonate
Catalog # APOSPC450482-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :325.095 g/mol
Storage Temperature:Ambient
CAS Number:1980085-62-8
Formula:C₉H₁₃BrF₄O₃
MDL Number:MFCD28334475