sec-Butyl 1H,1H,9H-perfluorononyl carbonate
Catalog # APOSPC450481-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:532.218 g/mol
Storage Temperature:Ambient
CAS Number:1923065-33-1
Formula:C₁₄H₁₂F₁₆O₃
MDL Number:MFCD28334453