sec-Butyl 1H,1H,7H-perfluorohexyl carbonate
Catalog # APOSPC450480-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:432.203 g/mol
Storage Temperature:Ambient
CAS Number:1980034-84-1
Formula:C₁₂H₁₂F₁₂O₃
MDL Number:MFCD28334452