sec-Butyl 1H,1H,2H,2H-perfluorohexyl carbonate
Catalog # APOSPC450479-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:364.205 g/mol
Storage Temperature:Ambient
CAS Number:1980034-90-9
Formula:C₁₁H₁₃F₉O₃
MDL Number:MFCD28334451