sec-Butyl 1H,1H-perfluorohexyl carbonate
Catalog # APOSPC450478-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:400.186 g/mol
Storage Temperature:Ambient
CAS Number:1980085-09-3
Formula:C₁₁H₁₁F₁₁O₃
MDL Number:MFCD28334450