sec-Butyl 4,4,5,5,5-pentafluoropentyl carbonate
Catalog # APOSPC450477-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:278.216 g/mol
Storage Temperature:Ambient
CAS Number:1923065-28-4
Formula:C₁₀H₁₅F₅O₃
MDL Number:MFCD28334449