sec-Butyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate
Catalog # APOSPC450475-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:300.171 g/mol
Storage Temperature:Ambient
CAS Number:1980038-46-7
Formula:C₉H₁₁F₇O₃
MDL Number:MFCD28334447