sec-Butyl 2,2,3,3-tetrafluoropropyl carbonate
Catalog # APOSPC450473-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:232.173 g/mol
Storage Temperature:Ambient
CAS Number:1980034-66-9
Formula:C₈H₁₂F₄O₃
MDL Number:MFCD28334445