sec-Butyl 2,2,3,3,3-pentafluoropropyl carbonate
Catalog # APOSPC450472-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:250.163 g/mol
Storage Temperature:Ambient
CAS Number:1923065-25-1
Formula:C₈H₁₁F₅O₃
MDL Number:MFCD28334444