Order Entry
Germany
Orders LinkContactUsLinkComponent
2-(2,2-Difluoroethoxy)ethanol

2-(2,2-Difluoroethoxy)ethanol

Flammable
Irritant
Supplier:   Apollo Scientific
2-(2,2-Difluoroethoxy)ethanol
Supplier:  Apollo Scientific
Certificates
2-(2,2-Difluoroethoxy)ethanol

3 Options Available Below

Product Details & Documents
  • CAS Number:853005-07-9
  • Formula:C₄H₈F₂O₂
  • MDL Number:MFCD19442036
  • Molecular Weight:126.102 g/mol
  • Storage Temperature:Refrigerator
  • UN:1993
  • ADR:3,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 79,60 €
    79,60 €
    1 * 1 g
       79,60 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 210,00 €
    210,00 €
    1 * 5 g
       210,00 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 956,00 €
    956,00 €
    1 * 25 g
       956,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.