4-(Trifluoromethoxy)indan-1-amine hydrochloride
Catalog # APOSPC449026-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:253.649 g/mol
Storage Temperature:Ambient
CAS Number:1083090-92-9
Formula:C₁₀H₁₀F₃NO·ClH
MDL Number:MFCD28975214