Order Entry
Germany
ContactUsLinkComponent
tert-Butyl 3-(trifluoromethyl)-2,5-dihydro-1H-pyrrole-1-carboxylate 95%
tert-Butyl 3-(trifluoromethyl)-2,5-dihydro-1H-pyrrole-1-carboxylate 95%
Catalog # APOSPC448210-100MG
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl 3-(trifluoromethyl)-2,5-dihydro-1H-pyrrole-1-carboxylate 95%
Catalog # APOSPC448210-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :237.219 g/mol
Storage Temperature:Ambient
CAS Number:693826-97-0
Formula:C₁₀H₁₄F₃NO₂
MDL Number:MFCD24468819