2-(3-Chloro-5-trifluoromethyl-pyridin-2-ylmethyl)-5-methyl-2H-pyrazole-3-carboxylic acid ethyl ester
Catalog # APOSPC446062-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:347.72 g/mol
Storage Temperature:Ambient
CAS Number:1351479-11-2
Formula:C₁₄H₁₃ClF₃N₃O₂
MDL Number:MFCD20526237